ENA282548243 Enamine

2-{[(thiophen-3-yl)methyl]amino}ethan-1-ol hydrochloride

  • MDL number MFCD07170344
  • Cas Number : 1158591-85-5
  • Molecular Weight : 193.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60753

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60753
Availability details for
Quantity
  • Availability
  • Add to Cart