ENA282548249 Enamine

3-(2-formylphenoxy)propanoic acid

  • MDL number MFCD11557064
  • Cas Number : 64603-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60759

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60759
Availability details for
Quantity
  • Availability
  • Add to Cart