ENA282548251 Enamine

3,4-dihydro-1H-2lambda6,3-benzothiazine-2,2,4-trione

  • MDL number MFCD01663228
  • Cas Number : 21784-53-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59509

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59509
Availability details for
Quantity
  • Availability
  • Add to Cart