ENA282548253 Enamine

N-[(piperidin-3-yl)methyl]propane-2-sulfonamide

  • MDL number MFCD12762778
  • Cas Number : 1178327-07-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60764

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60764
Availability details for
Quantity
  • Availability
  • Add to Cart