ENA282548255 Enamine

1-(4-ethylphenyl)-2-(morpholin-4-yl)ethan-1-one

  • MDL number MFCD12564747
  • Cas Number : 1152887-92-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60768

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60768
Availability details for
Quantity
  • Availability
  • Add to Cart