ENA282548261 Enamine

ethyl 4-(bromomethyl)-5,5-dimethyl-2-oxo-2,5-dihydrofuran-3-carboxylate

  • MDL number MFCD00698419
  • Cas Number : 90261-67-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59550

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59550
Availability details for
Quantity
  • Availability
  • Add to Cart