ENA282548262 Enamine

2-[(ethylcarbamoyl)amino]-6-fluorobenzoic acid

  • MDL number MFCD12171326
  • Cas Number : 1153395-12-0
  • Molecular Weight : 226.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60779

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60779
Availability details for
Quantity
  • Availability
  • Add to Cart