ENA282548264 Enamine

[(4-sulfamoylphenyl)carbamoyl]formic acid

  • MDL number MFCD00489479
  • Molecular Weight : 244.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-59574

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59574
Availability details for
Quantity
  • Availability
  • Add to Cart