ENA282548268 Enamine

5-methyl-N-(piperidin-4-yl)thiophene-2-carboxamide

  • MDL number MFCD11642757
  • Cas Number : 1098349-96-2
  • Molecular Weight : 224.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60787

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60787
Availability details for
Quantity
  • Availability
  • Add to Cart