ENA282548269 Enamine

6-(1-hydroxyethyl)-3,4-dihydro-2H-1,4-benzoxazin-3-one

  • MDL number MFCD12183778
  • Cas Number : 1154942-47-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60788

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60788
Availability details for
Quantity
  • Availability
  • Add to Cart