ENA282548272 Enamine

5-amino-1,3-dimethyl-2,3-dihydro-1H-1,3-benzodiazol-2-one

  • MDL number MFCD00462960
  • Cas Number : 53439-88-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148...150
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59698

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59698
Availability details for
Quantity
  • Availability
  • Add to Cart