ENA282548273 Enamine

5-chloro-2-(1H-imidazol-1-yl)aniline

  • MDL number MFCD04971013
  • Cas Number : 54705-92-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154...156
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60792

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60792
Availability details for
Quantity
  • Availability
  • Add to Cart