ENA282548278 Enamine

4-methyl-N-(piperidin-4-yl)-1,3-thiazole-5-carboxamide

  • MDL number MFCD11642758
  • Cas Number : 1094477-20-9
  • Molecular Weight : 225.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-60799

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60799
Availability details for
Quantity
  • Availability
  • Add to Cart