ENA282548280 Enamine

N'-hydroxy-2-(2-oxo-2,3-dihydro-1H-indol-3-yl)ethanimidamide

  • MDL number MFCD09046640
  • Cas Number : 926248-52-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 183...185
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60801

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60801
Availability details for
Quantity
  • Availability
  • Add to Cart