ENA282548282 Enamine

3-(1-hydroxyethyl)phenol

  • MDL number MFCD00238621
  • Cas Number : 2415-09-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-60803

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60803
Availability details for
Quantity
  • Availability
  • Add to Cart