ENA282548285 Enamine

2-(1H-imidazol-2-yl)piperidine

  • MDL number MFCD11111761
  • Cas Number : 933725-12-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144...146
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60807

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60807
Availability details for
Quantity
  • Availability
  • Add to Cart