ENA282548289 Enamine

N-(3-aminopropyl)ethane-1-sulfonamide

  • MDL number MFCD09941610
  • Cas Number : 1017047-84-5
  • Molecular Weight : 166.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-60811

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60811
Availability details for
Quantity
  • Availability
  • Add to Cart