ENA282548290 Enamine

(dicyclopropylmethyl)(methyl)amine

  • MDL number MFCD05864513
  • Cas Number : 884504-77-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60812

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60812
Availability details for
Quantity
  • Availability
  • Add to Cart