ENA282548297 Enamine

2-(4-nitrophenyl)-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD01936230
  • Cas Number : 17228-97-6
  • Molecular Weight : 250.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-60842

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60842
Availability details for
Quantity
  • Availability
  • Add to Cart