ENA282548298 Enamine

6-methyl-2-(pyrimidin-2-yl)-3,4-dihydropyrimidin-4-one

  • MDL number MFCD12093749
  • Cas Number : 1153999-07-5
  • Molecular Weight : 188.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59769

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59769
Availability details for
Quantity
  • Availability
  • Add to Cart