ENA282548301 Enamine

4-tert-butyl-1,2,5-oxadiazol-3-amine

  • MDL number MFCD11184683
  • Cas Number : 159013-87-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-43671

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43671
Availability details for
Quantity
  • Availability
  • Add to Cart