ENA282548306 Enamine

4-methoxy-3-phenylbenzoic acid

  • MDL number MFCD09042442
  • Cas Number : 4477-64-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-60851

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60851
Availability details for
Quantity
  • Availability
  • Add to Cart