ENA282548307 Enamine

1-(2-chlorophenyl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD10689969
  • Cas Number : 1152533-76-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-60852

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60852
Availability details for
Quantity
  • Availability
  • Add to Cart