ENA282548308 Enamine

2-aminopropanamide

  • MDL number MFCD07841687
  • Cas Number : 4726-84-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 52...54
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60853

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-60853
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart