ENA282548316 Enamine

morpholine-2-carboxamide

  • MDL number MFCD15209643
  • Cas Number : 135072-13-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158...160
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60862

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-60862
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart