ENA282548317 Enamine

4-(cyclopentanesulfonyl)benzoic acid

  • MDL number MFCD09940123
  • Cas Number : 860697-74-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60863

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60863
Availability details for
Quantity
  • Availability
  • Add to Cart