ENA282548318 Enamine

diethyl[(piperidin-3-yl)methyl]amine dihydrochloride

  • MDL number MFCD13561485
  • Cas Number : 1220021-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59980

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59980
Availability details for
Quantity
  • Availability
  • Add to Cart