ENA282548323 Enamine

4-bromo-N-[(4-methylphenyl)methyl]-N-propyl-1H-pyrrole-2-carboxamide

  • MDL number MFCD15209648
  • Cas Number : 1183918-20-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60871

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60871
Availability details for
Quantity
  • Availability
  • Add to Cart