ENA282548329 Enamine

2-[(dimethylamino)methyl]cyclohexan-1-ol

  • MDL number MFCD01037909
  • Cas Number : 17589-70-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60880

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60880
Availability details for
Quantity
  • Availability
  • Add to Cart