ENA282548333 Enamine

N-(2-methylphenyl)piperidin-4-amine

  • MDL number MFCD00101443
  • Cas Number : 1183200-00-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-60884

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60884
Availability details for
Quantity
  • Availability
  • Add to Cart