ENA282548342 Enamine

2-(3-methyl-1H-indol-1-yl)acetic acid

  • MDL number MFCD11641408
  • Cas Number : 57710-28-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175...177
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60906

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60906
Availability details for
Quantity
  • Availability
  • Add to Cart