ENA282548353 Enamine

N1-(pyridin-4-yl)propane-1,3-diamine

  • MDL number MFCD09932459
  • Cas Number : 1016501-09-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60918

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60918
Availability details for
Quantity
  • Availability
  • Add to Cart