ENA282548357 Enamine

1-[1-(4-chlorophenyl)-5-methyl-1H-1,2,3-triazol-4-yl]ethan-1-one

  • MDL number MFCD03236301
  • Cas Number : 33821-38-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60923

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60923
Availability details for
Quantity
  • Availability
  • Add to Cart