ENA282548359 Enamine

2-(pyridin-3-yloxy)benzoic acid

  • MDL number MFCD11182950
  • Cas Number : 1094325-06-0
  • Molecular Weight : 215.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-60925

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60925
Availability details for
Quantity
  • Availability
  • Add to Cart