ENA282548361 Enamine

2-bromo-3-(chlorosulfonyl)-5-fluorobenzoic acid

  • MDL number MFCD09043932
  • Cas Number : 926194-65-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 186...188
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60929

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60929
Availability details for
Quantity
  • Availability
  • Add to Cart