ENA282548362 Enamine

1-[2-(morpholin-4-yl)ethyl]-1H-indol-5-amine

  • MDL number MFCD11213931
  • Cas Number : 1094544-00-9
  • Molecular Weight : 245.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60930

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60930
Availability details for
Quantity
  • Availability
  • Add to Cart