ENA282548363 Enamine

1-[(3-nitrophenyl)carbonyl]piperazine

  • MDL number MFCD04116576
  • Cas Number : 341529-34-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145...147
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60931

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60931
Availability details for
Quantity
  • Availability
  • Add to Cart