ENA282548364 Enamine

3-[(4-methylphenyl)methyl]piperidine

  • MDL number MFCD05863597
  • Cas Number : 136421-81-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60933

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60933
Availability details for
Quantity
  • Availability
  • Add to Cart