ENA282548368 Enamine

2,4-dinitrobenzene-1-sulfonamide

  • MDL number MFCD09733471
  • Cas Number : 73901-01-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149...151
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60073

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60073
Availability details for
Quantity
  • Availability
  • Add to Cart