ENA282548371 Enamine

2-methanesulfonylcyclohexan-1-one

  • MDL number MFCD15209655
  • Cas Number : 16096-71-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-60942

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60942
Availability details for
Quantity
  • Availability
  • Add to Cart