ENA282548375 Enamine

5-phenyl-1-(2,2,2-trifluoroethyl)-1H-imidazole-2-thiol

  • MDL number MFCD11182662
  • Cas Number : 1094354-75-2
  • Molecular Weight : 258.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-phenyl-1-(2,2,2-trifluoroethyl)-1H-imidazole-2-thiol
Inchi Code 1S/C11H9F3N2S/c12-11(13,14)7-16-9(6-15-10(16)17)8-4-2-1-3-5-8/h1-6H,7H2,(H,15,17)
Inchi Key VGMDTVDXUFYCPS-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-60953

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60953
Availability details for
Quantity
  • Availability
  • Add to Cart