ENA282548376 Enamine

4-(4-methyl-1,3-thiazol-2-yl)benzoic acid

  • MDL number MFCD11609891
  • Cas Number : 933709-38-7
  • Molecular Weight : 219.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-60954

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60954
Availability details for
Quantity
  • Availability
  • Add to Cart