ENA282548381 Enamine

4-[(propane-2-sulfonyl)methyl]aniline

  • MDL number MFCD12801280
  • Cas Number : 635702-61-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60959

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60959
Availability details for
Quantity
  • Availability
  • Add to Cart