ENA282548389 Enamine

1-(4-fluorophenyl)-2-(1H-imidazol-1-yl)ethan-1-one

  • MDL number MFCD03826462
  • Cas Number : 24155-33-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143-145
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-60967

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60967
Availability details for
Quantity
  • Availability
  • Add to Cart