ENA282548394 Enamine

1-(2,3-dihydro-1,4-benzodioxine-6-sulfonyl)-1,4-diazepane

  • MDL number MFCD11583412
  • Cas Number : 1082556-61-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60974

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60974
Availability details for
Quantity
  • Availability
  • Add to Cart