ENA282548395 Enamine

methyl 2-(4-tert-butyl-1,3-thiazol-2-yl)acetate

  • MDL number MFCD14630525
  • Cas Number : 1221724-38-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-60975

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60975
Availability details for
Quantity
  • Availability
  • Add to Cart