ENA282548397 Enamine

3-cyclobutyl-3-oxopropanenitrile

  • MDL number MFCD07787182
  • Cas Number : 118431-89-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. -20

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60977
Availability details for
Quantity
  • Availability
  • Add to Cart