ENA282548399 Enamine

1,3-dimethylpiperidin-4-amine

  • MDL number MFCD09035302
  • Cas Number : 900642-45-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60979

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60979
Availability details for
Quantity
  • Availability
  • Add to Cart