ENA282548401 Enamine

[2-(dimethylamino)ethyl][(oxolan-2-yl)methyl]amine

  • MDL number MFCD08059858
  • Cas Number : 915921-74-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60981

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60981
Availability details for
Quantity
  • Availability
  • Add to Cart