ENA282548406 Enamine

3-amino-1-{4H,5H,6H,7H-thieno[3,2-c]pyridin-5-yl}propan-1-one

  • MDL number MFCD09938104
  • Cas Number : 1017049-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60986

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60986
Availability details for
Quantity
  • Availability
  • Add to Cart